Assay Details
Gene Family ID: | MIPF0000911, mir-2797 |
Mature miRNA Sequence: | UUAAGUAGAUUCUAUUUGGCUA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Manduca sexta | mse-miR-2797a | MIMAT0024499 |
Stem-loop Details
Manduca sexta
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | mse-miR-2797a |
|---|---|
| miRBase Accession Number: | MI0021049 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUAAGUAGAUUCUAUUUGGCUA |
| Species: | Manduca sexta |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000911, mir-2797 |
Mature miRNA Sequence: | UUAAGUAGAUUCUAUUUGGCUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Manduca sexta | mse-miR-2797a | MIMAT0024499 |