Assay Details
Mature miRNA Sequence: | ACCCUAUCAAUAUUGCCUCUGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Chrysemys picta | cpi-miR-454-5p | MIMAT0037885 |
| Salmo salar | ssa-miR-454-5p | MIMAT0032641 |
Stem-loop Details
Chrysemys picta
Salmo salar

Genomic Map