Assay Details
Gene Family ID: | MIPF0000098, mir-95 |
Mature miRNA Sequence: | CUCAGUAAAUGUUUAUUGGAU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Goat | chi-miR-545-5p | MIMAT0036277 |
Stem-loop Details
Goat
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | chi-miR-545-5p |
|---|---|
| miRBase Accession Number: | MI0030822 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUCAGUAAAUGUUUAUUGGAU |
| Species: | Goat |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000098, mir-95 |
Mature miRNA Sequence: | CUCAGUAAAUGUUUAUUGGAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Goat | chi-miR-545-5p | MIMAT0036277 |