Assay Details
Gene Family ID: | MIPF0000019, mir-8 |
Mature miRNA Sequence: | GUCUUACCAGACAAAUUUAGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ophiophagus hannah | oha-miR-429-5p | MIMAT0036923 |
Stem-loop Details
Ophiophagus hannah
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | oha-miR-429-5p |
|---|---|
| miRBase Accession Number: | MI0031469 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GUCUUACCAGACAAAUUUAGA |
| Species: | Ophiophagus hannah |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000019, mir-8 |
Mature miRNA Sequence: | GUCUUACCAGACAAAUUUAGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ophiophagus hannah | oha-miR-429-5p | MIMAT0036923 |