Assay Details
Mature miRNA Sequence: | UAAGGUGCAUCUAGUGCAGAUA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Astatotilapia burtoni | abu-miR-18a | MIMAT0042137 |
| Ateles geoffroyi | age-miR-18 | MIMAT0002674 |
| Bovine | bta-miR-18a | MIMAT0003526 |
| Chicken | gga-miR-18a-5p | MIMAT0001113 |
| Columba livia | cli-miR-18a-5p | MIMAT0038423 |
| Dog | cfa-miR-18a | MIMAT0009832 |
| View More Rows | ||
| Fugu rubripes | fru-miR-18 | MIMAT0002918 |
| Gadus morhua | gmo-miR-18a-5p | MIMAT0044321 |
| Gorilla gorilla | ggo-miR-18a | MIMAT0002660 |
| Lagothrix lagotricha | lla-miR-18 | MIMAT0002716 |
| Lemur catta | lca-miR-18 | MIMAT0002667 |
| Macaca nemestrina | mne-miR-18 | MIMAT0002723 |
| Metriaclima zebra | mze-miR-18a | MIMAT0042362 |
| Monodelphis domestica | mdo-miR-18a-5p | MIMAT0004167 |
| Neolamprologus brichardi | nbr-miR-18a | MIMAT0042537 |
| Oreochromis niloticus | oni-miR-18a | MIMAT0042726 |
| Pan paniscus | ppa-miR-18 | MIMAT0002681 |
| Pan troglodytes | ptr-miR-18a | MIMAT0002695 |
| Pig | ssc-miR-18a | MIMAT0002161 |
| Pongo pygmaeus | ppy-miR-18a | MIMAT0002688 |
| Pteropus alecto | pal-miR-18a-5p | MIMAT0040222 |
| Pundamilia nyererei | pny-miR-18a | MIMAT0042923 |
| Rhesus monkey | mml-miR-18a-5p | MIMAT0002702 |
| Saguinus labiatus | sla-miR-18 | MIMAT0002709 |
| Salmo salar | ssa-miR-18b-5p | MIMAT0032434 |
| Taeniopygia guttata | tgu-miR-18a | MIMAT0014587 |
| Tetraodon nigroviridis | tni-miR-18 | MIMAT0002919 |
| Zebrafish | dre-miR-18a | MIMAT0001779 |
Stem-loop Details
Astatotilapia burtoni
Ateles geoffroyi
Bovine
Chicken
Columba livia
Dog
Fugu rubripes
Gadus morhua
Gorilla gorilla
Lagothrix lagotricha
Lemur catta
Macaca nemestrina
Metriaclima zebra
Monodelphis domestica
Neolamprologus brichardi
Oreochromis niloticus
Pan paniscus
Pan troglodytes
Pig
Pongo pygmaeus
Pteropus alecto
Pundamilia nyererei
Rhesus monkey
Saguinus labiatus
Salmo salar
Taeniopygia guttata
Tetraodon nigroviridis
Zebrafish
More Information
Related Products
miRBase Alias:
Gallus gallus: gga-miR-18a (v19)
Gorilla gorilla adenovirus B7: ggo-miR-18 (v18)
Macaca mulatta: mml-miR-18 (v18)
Macaca mulatta: mml-miR-18a (v19)
Monodelphis domestica: mdo-miR-18 (v19)
Pongo pygmaeus: ppy-miR-18 (v18)
Staphylococcus schleiferi: ssc-miR-18 (v16)
Gorilla gorilla adenovirus B7: ggo-miR-18 (v18)
Macaca mulatta: mml-miR-18 (v18)
Macaca mulatta: mml-miR-18a (v19)
Monodelphis domestica: mdo-miR-18 (v19)
Pongo pygmaeus: ppy-miR-18 (v18)
Staphylococcus schleiferi: ssc-miR-18 (v16)

Genomic Map