Assay Details
Mature miRNA Sequence: | UAGUGCAAUAUUGCUUAUAGGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Astatotilapia burtoni | abu-miR-454b | MIMAT0042247 |
Dog | cfa-miR-454 | MIMAT0009927 |
Pongo pygmaeus | ppy-miR-454 | MIMAT0024367 |
Zebrafish | dre-miR-454b | MIMAT0001878 |
Stem-loop Details
Astatotilapia burtoni
Dog
Pongo pygmaeus
Zebrafish
