Assay Details
Gene Family ID: | MIPF0000002, let-7 |
Mature miRNA Sequence: | UGAGGUAGUAGUUUGUACAGUU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-let-7g-5p | MIMAT0000414 |
Mouse | mmu-let-7g-5p | MIMAT0000121 |
Rat | rno-let-7g-5p | MIMAT0035719 |
Alligator mississippiensis | ami-let-7g-5p | MIMAT0038014 |
Anolis carolinensis | aca-let-7g | MIMAT0021706 |
Bovine | bta-let-7g | MIMAT0003838 |
View More Rows | ||
Callithrix jacchus | cja-let-7g | MIMAT0039326 |
Chrysemys picta | cpi-let-7g-5p | MIMAT0037608 |
Columba livia | cli-let-7g-5p | MIMAT0038387 |
Cricetulus griseus | cgr-let-7g-5p | MIMAT0023722 |
Dasypus novemcinctus | dno-let-7g-5p | MIMAT0047485 |
Daubentonia madagascariensis | dma-let-7g | MIMAT0049290 |
Dog | cfa-let-7g | MIMAT0006637 |
Goat | chi-let-7g-5p | MIMAT0035892 |
Guinea pig | cpo-let-7g-5p | MIMAT0046831 |
Horse | eca-let-7g | MIMAT0013075 |
Microcebus murinus | mmr-let-7g | MIMAT0049151 |
Nomascus leucogenys | nle-let-7g | MIMAT0049456 |
Ophiophagus hannah | oha-let-7g-5p | MIMAT0036640 |
Ornithorhynchus anatinus | oan-let-7g-5p | MIMAT0007038 |
Otolemur garnettii | oga-let-7g | MIMAT0049649 |
Pan paniscus | ppa-let-7g | MIMAT0049075 |
Pan troglodytes | ptr-let-7g | MIMAT0007942 |
Papio hamadryas | pha-let-7g | MIMAT0049574 |
Pig | ssc-let-7g | MIMAT0013867 |
Pongo pygmaeus | ppy-let-7g | MIMAT0015727 |
Pteropus alecto | pal-let-7g-5p | MIMAT0039774 |
Python bivittatus | pbv-let-7g-5p | MIMAT0038808 |
Rabbit | ocu-let-7g-5p | MIMAT0048087 |
Rhesus monkey | mml-let-7g-5p | MIMAT0006157 |
Taeniopygia guttata | tgu-let-7g-5p | MIMAT0014528 |
Tupaia chinensis | tch-let-7g-5p | MIMAT0036519 |
Stem-loop Details
Human
Stem-loop ID | hsa-let-7g |
Stem-loop Accession # | MI0000433 |
Stem-loop Sequence | AGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCA |
Chromosome Location | Chr. 3 - 52268278 - 52268361 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000414
miRBase ID: hsa-let-7g-5p Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: Chr. 3 - 52268278 - 52268361 [-] on Build GRCh38 |
Stem-loop ID | mmu-let-7g |
Stem-loop Accession # | MI0000137 |
Stem-loop Sequence | CCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCAGG |
Chromosome Location | Chr. 9 - 106178840 - 106178927 [+] on Build GRCm38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000121
miRBase ID: mmu-let-7g-5p Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: Chr. 9 - 106178840 - 106178927 [+] on Build GRCm38 |
Stem-loop ID | rno-let-7g |
Stem-loop Accession # | MI0030347 |
Stem-loop Sequence | CCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCAGG |
Chromosome Location | Chr. 8 - 114874457 - 114874544 [+] on Build Rnor_6.0 |
Mature MicroRNA |
miRBase Accession #: MIMAT0035719
miRBase ID: rno-let-7g-5p Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: Chr. 8 - 114874457 - 114874544 [+] on Build Rnor_6.0 |
Stem-loop ID | ami-let-7g |
Stem-loop Accession # | MI0029650 |
Stem-loop Sequence | GCCUGAUUCCAAGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUUAACUGUACAGGCCACUGCCUUGCCUGGGACAG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0038014
miRBase ID: ami-let-7g-5p Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | aca-let-7g |
Stem-loop Accession # | MI0018709 |
Stem-loop Sequence | UUGCCUGAUUCCGGGUUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCUGGGGCAGAGCUUU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0021706
miRBase ID: aca-let-7g Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | bta-let-7g |
Stem-loop Accession # | MI0005051 |
Stem-loop Sequence | AGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0003838
miRBase ID: bta-let-7g Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | cja-let-7g |
Stem-loop Accession # | MI0031835 |
Stem-loop Sequence | GCUCCGUUUCCUUUUGCCUGAUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCAGGAACAGUGCGCCAGCUGCCAAAUGGGGC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0039326
miRBase ID: cja-let-7g Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | cpi-let-7g |
Stem-loop Accession # | MI0029382 |
Stem-loop Sequence | UCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCUGGGA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0037608
miRBase ID: cpi-let-7g-5p Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | cli-let-7g |
Stem-loop Accession # | MI0029891 |
Stem-loop Sequence | UUCUUGCCUGAUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCUGGGACAGA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0038387
miRBase ID: cli-let-7g-5p Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | cgr-let-7g |
Stem-loop Accession # | MI0020373 |
Stem-loop Sequence | AUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCAGGAACAGC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0023722
miRBase ID: cgr-let-7g-5p Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | dno-let-7g |
Stem-loop Accession # | MI0038893 |
Stem-loop Sequence | UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0047485
miRBase ID: dno-let-7g-5p Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | dma-let-7g |
Stem-loop Accession # | MI0040023 |
Stem-loop Sequence | UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049290
miRBase ID: dma-let-7g Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | cfa-let-7g |
Stem-loop Accession # | MI0008036 |
Stem-loop Sequence | UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0006637
miRBase ID: cfa-let-7g Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | chi-let-7g |
Stem-loop Accession # | MI0030585 |
Stem-loop Sequence | CCUUUUGCCUGAUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCGGGAACAG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0035892
miRBase ID: chi-let-7g-5p Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | cpo-let-7g |
Stem-loop Accession # | MI0038549 |
Stem-loop Sequence | UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUAUAGGAGAUAACUGUACAGGCCACUGCCUUGCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0046831
miRBase ID: cpo-let-7g-5p Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | eca-let-7g |
Stem-loop Accession # | MI0012820 |
Stem-loop Sequence | CCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCAGG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0013075
miRBase ID: eca-let-7g Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | mmr-let-7g |
Stem-loop Accession # | MI0039866 |
Stem-loop Sequence | UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUAUAGGAGAUAACUGUACAGGCCACUGCCUUGCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049151
miRBase ID: mmr-let-7g Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | nle-let-7g |
Stem-loop Accession # | MI0040208 |
Stem-loop Sequence | UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049456
miRBase ID: nle-let-7g Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | oha-let-7g |
Stem-loop Accession # | MI0031309 |
Stem-loop Sequence | GGUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCUGGGGCAG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0036640
miRBase ID: oha-let-7g-5p Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | oan-let-7g |
Stem-loop Accession # | MI0006809 |
Stem-loop Sequence | UCCAGGCUGAGGUAGUAGUUUGUACAGUUUCAGGGUCUACAAUACCACCCGGUAUAGGAGAUAACUGUACAGGCCACUGCCUUGCCAGGGA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0007038
miRBase ID: oan-let-7g-5p Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | oga-let-7g |
Stem-loop Accession # | MI0040432 |
Stem-loop Sequence | UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUGUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049649
miRBase ID: oga-let-7g Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | ppa-let-7g |
Stem-loop Accession # | MI0039783 |
Stem-loop Sequence | UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049075
miRBase ID: ppa-let-7g Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | ptr-let-7g |
Stem-loop Accession # | MI0008406 |
Stem-loop Sequence | AGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0007942
miRBase ID: ptr-let-7g Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | pha-let-7g |
Stem-loop Accession # | MI0040349 |
Stem-loop Sequence | UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049574
miRBase ID: pha-let-7g Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | ssc-let-7g |
Stem-loop Accession # | MI0013087 |
Stem-loop Sequence | GGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0013867
miRBase ID: ssc-let-7g Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | ppy-let-7g |
Stem-loop Accession # | MI0014788 |
Stem-loop Sequence | AGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0015727
miRBase ID: ppy-let-7g Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | pal-let-7g |
Stem-loop Accession # | MI0032321 |
Stem-loop Sequence | UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0039774
miRBase ID: pal-let-7g-5p Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | pbv-let-7g |
Stem-loop Accession # | MI0030141 |
Stem-loop Sequence | GUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUACGAUACCGCCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCUGGGGC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0038808
miRBase ID: pbv-let-7g-5p Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | ocu-let-7g |
Stem-loop Accession # | MI0039213 |
Stem-loop Sequence | UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0048087
miRBase ID: ocu-let-7g-5p Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | mml-let-7g |
Stem-loop Accession # | MI0007579 |
Stem-loop Sequence | AGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0006157
miRBase ID: mml-let-7g-5p Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | tgu-let-7g-1 |
Stem-loop Accession # | MI0013740 |
Stem-loop Sequence | GAUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCUGG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0014528
miRBase ID: tgu-let-7g-5p Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | tgu-let-7g-2 |
Stem-loop Accession # | MI0013741 |
Stem-loop Sequence | GAUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCUGG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0014528
miRBase ID: tgu-let-7g-5p Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |
Stem-loop ID | tch-let-7g |
Stem-loop Accession # | MI0031177 |
Stem-loop Sequence | UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0036519
miRBase ID: tch-let-7g-5p Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU Chromosome Location: NA |

More Information
Related Products








miRBase Alias:
Human: hsa-let-7g (v17)
Mouse: mmu-let-7g (v17)
Macaca mulatta: mml-let-7g (v19)
Ochrobactrum anthropi: oan-let-7g (v18)
Trabulsiella guamensis ATCC 49490: tgu-let-7g (v19)
Mouse: mmu-let-7g (v17)
Macaca mulatta: mml-let-7g (v19)
Ochrobactrum anthropi: oan-let-7g (v18)
Trabulsiella guamensis ATCC 49490: tgu-let-7g (v19)
