Assay Details
Mature miRNA Sequence: | UGGUGAAAAAUAUGAAAAAUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Physcomitrella patens | ppt-miR1046-3p | MIMAT0005160 |
Stem-loop Details
Physcomitrella patens
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ppt-miR1046-3p |
|---|---|
| miRBase Accession Number: | MI0006005 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGUGAAAAAUAUGAAAAAUC |
| Species: | Physcomitrella patens |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | UGGUGAAAAAUAUGAAAAAUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Physcomitrella patens | ppt-miR1046-3p | MIMAT0005160 |