Assay Details
Gene Family ID: | MIPF0000098, mir-95 |
Mature miRNA Sequence: | CAAGUCUUAUUUGAGCACCUGUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1264 | MIMAT0005791 |
| Callithrix jacchus | cja-miR-1264 | MIMAT0039604 |
| Horse | eca-miR-1264 | MIMAT0013195 |
| Pan troglodytes | ptr-miR-1264 | MIMAT0007988 |
| Pongo pygmaeus | ppy-miR-1264 | MIMAT0016188 |
Stem-loop Details
Human
Callithrix jacchus
Horse
Pan troglodytes
Pongo pygmaeus

Genomic Map