Assay Details
Gene Family ID: | MIPF0000531, mir-1306 |
Mature miRNA Sequence: | CCACCUCCCCUGCAAACGUCC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-miR-1306 | MIMAT0009974 |
| Columba livia | cli-miR-1306-5p | MIMAT0038670 |
| Cricetulus griseus | cgr-miR-1306-5p | MIMAT0023752 |
| Dog | cfa-miR-1306 | MIMAT0006661 |
| Goat | chi-miR-1306-5p | MIMAT0035939 |
| Monodelphis domestica | mdo-miR-1306-5p | MIMAT0050231 |
| View More Rows | ||
| Oryzias latipes | ola-miR-1306 | MIMAT0022590 |
| Rhesus monkey | mml-miR-1306-5p | MIMAT0028240 |
Stem-loop Details
Bovine
Columba livia
Cricetulus griseus
Dog
Goat
Monodelphis domestica
Oryzias latipes
Rhesus monkey

Genomic Map