Assay Details
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | UAUGUAAUAUGGUCCACGUCUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Horse | eca-miR-380 | MIMAT0013148 |
| Rhesus monkey | mml-miR-380-3p | MIMAT0006309 |
Stem-loop Details
Horse
Rhesus monkey

Genomic Map