Assay Details
Gene Family ID: | MIPF0000585, mir-1237 |
Mature miRNA Sequence: | UCCUUCUGCUCCGUCCCCCAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1237-3p | MIMAT0005592 |
| Pan troglodytes | ptr-miR-1237 | MIMAT0007969 |
Stem-loop Details
Human
Pan troglodytes

Genomic Map