Assay Details
Gene Family ID: | MIPF0000574, mir-1420 |
Mature miRNA Sequence: | AAAGUGCAACCAGAUUGGAUGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ornithorhynchus anatinus | oan-miR-1347 | MIMAT0006930 |
Stem-loop Details
Ornithorhynchus anatinus

Genomic Map