Assay Details
Mature miRNA Sequence: | CAAAUGCACGCAAAUGAAGAUGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Nematostella vectensis | nve-miR-2049-5p | MIMAT0009822 |
Stem-loop Details
Nematostella vectensis
More Information
Related Products
miRBase Alias:
Nematostella vectensis: nve-miR-2049 (v18)

Genomic Map