Assay Details
Gene Family ID: | MIPF0000725, mir-1287 |
Mature miRNA Sequence: | UGCUGGAUCAGUGGUUUGAGUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-miR-1287 | MIMAT0009971 |
| Pongo pygmaeus | ppy-miR-1287 | MIMAT0016210 |
Stem-loop Details
Bovine
Pongo pygmaeus

Genomic Map