Assay Details
Mature miRNA Sequence: | CUAUCUACUUAAAAUGUGCCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis elegans | cel-miR-2216-3p | MIMAT0011448 |
Stem-loop Details
Caenorhabditis elegans
More Information
Related Products
miRBase Alias:
Caenorhabditis elegans: cel-miR-2216* (v17)

Genomic Map