Assay Details
Mature miRNA Sequence: | UAGUAGGUUGUGUUGUUUAGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-miR-2354 | MIMAT0011890 |
Stem-loop Details
Bovine
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | bta-miR-2354 |
|---|---|
| miRBase Accession Number: | MI0011383 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAGUAGGUUGUGUUGUUUAGU |
| Species: | Bovine |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | UAGUAGGUUGUGUUGUUUAGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-miR-2354 | MIMAT0011890 |