Assay Details
Gene Family ID: | MIPF0000787, mir-2209 |
Mature miRNA Sequence: | AGAGAUCAGCGGUUACACUACA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis elegans | cel-miR-2209a-3p | MIMAT0011432 |
Stem-loop Details
Caenorhabditis elegans
More Information
Related Products
miRBase Alias:
Caenorhabditis elegans: cel-miR-2209a (v17)

Genomic Map