Assay Details
Gene Family ID: | MIPF0000937, mir-307as |
Mature miRNA Sequence: | UCACUCAAGGAGGUUGUGAUGG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-307b-5p | MIMAT0012194 |
Stem-loop Details
Drosophila melanogaster
More Information
Related Products
miRBase Alias:
Drosophila melanogaster: dme-miR-307-as* (v16)

Genomic Map