Assay Details
Gene Family ID: | MIPF0000610, mir-1290 |
Mature miRNA Sequence: | UGGAUUUUUGGAUCAGAGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pongo pygmaeus | ppy-miR-1290 | MIMAT0016212 |
Stem-loop Details
Pongo pygmaeus
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ppy-miR-1290 |
|---|---|
| miRBase Accession Number: | MI0015246 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGAUUUUUGGAUCAGAGA |
| Species: | Pongo pygmaeus |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000610, mir-1290 |
Mature miRNA Sequence: | UGGAUUUUUGGAUCAGAGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pongo pygmaeus | ppy-miR-1290 | MIMAT0016212 |