Assay Details
Gene Family ID: | MIPF0000114, mir-375 |
Mature miRNA Sequence: | ACUUGGGCCAAGGGAAUGCAAACU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-375-5p | MIMAT0020853 |
| Drosophila simulans | dsi-miR-375-5p | MIMAT0012489 |
| Drosophila virilis | dvi-miR-375-5p | MIMAT0035568 |
Stem-loop Details
Drosophila melanogaster
Drosophila simulans
Drosophila virilis
More Information
Related Products
miRBase Alias:
Drosophila simulans: dsi-miR-375* (v18)

Genomic Map