Assay Details
Gene Family ID: | MIPF0000715, mir-1175 |
Mature miRNA Sequence: | AAGUGGAGGUGUGAUCUCUUCA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bombyx mori | bmo-miR-1175-5p | MIMAT0013612 |
Heliconius melpomene | hme-miR-1175 | MIMAT0024935 |
Stem-loop Details
Bombyx mori
Heliconius melpomene
