Assay Details
Gene Family ID: | MIPF0000827, mir-2778 |
Mature miRNA Sequence: | GUUAUUUUGCAUAUCCUGCA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bombyx mori | bmo-miR-2778a-4-5p | MIMAT0015306 |
Stem-loop Details
Bombyx mori
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | bmo-miR-2778a-4-5p |
|---|---|
| miRBase Accession Number: | MI0012353 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GUUAUUUUGCAUAUCCUGCA |
| Species: | Bombyx mori |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000827, mir-2778 |
Mature miRNA Sequence: | GUUAUUUUGCAUAUCCUGCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bombyx mori | bmo-miR-2778a-4-5p | MIMAT0015306 |