Assay Details
Mature miRNA Sequence: | UCUCCCCAUUCCUUGUGAUUGCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pristionchus pacificus | ppc-miR-2247 | MIMAT0011735 |
Stem-loop Details
Pristionchus pacificus
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ppc-miR-2247 |
|---|---|
| miRBase Accession Number: | MI0011243 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCUCCCCAUUCCUUGUGAUUGCU |
| Species: | Pristionchus pacificus |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | UCUCCCCAUUCCUUGUGAUUGCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pristionchus pacificus | ppc-miR-2247 | MIMAT0011735 |