Assay Details
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | CCCUUCCUGGUAUUUGAAGACG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Sheep | oar-miR-1197-5p | MIMAT0019257 |
Stem-loop Details
Sheep
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | oar-miR-1197-5p |
|---|---|
| miRBase Accession Number: | MI0016922 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCCUUCCUGGUAUUUGAAGACG |
| Species: | Sheep |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | CCCUUCCUGGUAUUUGAAGACG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Sheep | oar-miR-1197-5p | MIMAT0019257 |