Assay Details
Gene Family ID: | MIPF0000141, mir-315 |
Mature miRNA Sequence: | AUUUGAUUGUUGCUCAGAAAGCCG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Petromyzon marinus | pma-miR-315 | MIMAT0019539 |
Stem-loop Details
Petromyzon marinus
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | pma-miR-315 |
|---|---|
| miRBase Accession Number: | MI0017124 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AUUUGAUUGUUGCUCAGAAAGCCG |
| Species: | Petromyzon marinus |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000141, mir-315 |
Mature miRNA Sequence: | AUUUGAUUGUUGCUCAGAAAGCCG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Petromyzon marinus | pma-miR-315 | MIMAT0019539 |