Assay Details
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | UAGGACACAUGGUCUACUUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Gorilla gorilla | ggo-miR-1197 | MIMAT0024245 |
Stem-loop Details
Gorilla gorilla
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ggo-miR-1197 |
|---|---|
| miRBase Accession Number: | MI0020792 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAGGACACAUGGUCUACUUC |
| Species: | Gorilla gorilla |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | UAGGACACAUGGUCUACUUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Gorilla gorilla | ggo-miR-1197 | MIMAT0024245 |