Assay Details
Gene Family ID: | MIPF0000531, mir-1306 |
Mature miRNA Sequence: | ACCACCUCCCCUGCAAACGUCCAGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Chicken | gga-miR-1306-5p | MIMAT0026763 |
Stem-loop Details
Chicken
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | gga-miR-1306-5p |
|---|---|
| miRBase Accession Number: | MI0007057 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACCACCUCCCCUGCAAACGUCCAGU |
| Species: | Chicken |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000531, mir-1306 |
Mature miRNA Sequence: | ACCACCUCCCCUGCAAACGUCCAGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Chicken | gga-miR-1306-5p | MIMAT0026763 |