Assay Details
Mature miRNA Sequence: | CUACAGCACGAAACUGCCUUAA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Chicken | gga-miR-196-2-3p | MIMAT0026504 |
| Columba livia | cli-miR-196-3-3p | MIMAT0038572 |
Stem-loop Details
Chicken
Columba livia
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | cli-miR-196-3-3p |
|---|---|
| miRBase Accession Number: | MI0030002 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUACAGCACGAAACUGCCUUAA |
| Species: | Chicken, Columba livia |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | CUACAGCACGAAACUGCCUUAA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Chicken | gga-miR-196-2-3p | MIMAT0026504 |
| Columba livia | cli-miR-196-3-3p | MIMAT0038572 |