Assay Details
Gene Family ID: | MIPF0000130, mir-506 |
Mature miRNA Sequence: | AAUGUGUAGCAAAAGACAGAAU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rhesus monkey | mml-miR-511-3p | MIMAT0026893 |
Stem-loop Details
Rhesus monkey

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | mml-miR-511-3p |
---|---|
miRBase Accession Number: | MI0007780 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AAUGUGUAGCAAAAGACAGAAU |
Species: | Rhesus monkey |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000130, mir-506 |
Mature miRNA Sequence: | AAUGUGUAGCAAAAGACAGAAU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rhesus monkey | mml-miR-511-3p | MIMAT0026893 |