Assay Details
Mature miRNA Sequence: | ACCCUAUCAAUAUUGCCUCUGCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Alligator mississippiensis | ami-miR-454-5p | MIMAT0038285 |
| Columba livia | cli-miR-454-5p | MIMAT0038644 |
| Taeniopygia guttata | tgu-miR-454-5p | MIMAT0027011 |
Stem-loop Details
Alligator mississippiensis
Columba livia
Taeniopygia guttata

Genomic Map