Assay Details
Mature miRNA Sequence: | UAGUGCAAUAUUGCUUAUAGGGUCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Alligator mississippiensis | ami-miR-454-3p | MIMAT0038286 |
| Columba livia | cli-miR-454-3p | MIMAT0038645 |
| Oreochromis niloticus | oni-miR-454b | MIMAT0043039 |
| Salmo salar | ssa-miR-454-3p | MIMAT0032642 |
Stem-loop Details
Alligator mississippiensis
Columba livia
Oreochromis niloticus
Salmo salar

Genomic Map