Assay Details
Gene Family ID: | MIPF0000168, mir-378 |
Mature miRNA Sequence: | CACUGGACUUGGAGUCAGAAGGCC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Eptesicus fuscus | efu-miR-378 | MIMAT0034813 |
Stem-loop Details
Eptesicus fuscus
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | efu-miR-378 |
|---|---|
| miRBase Accession Number: | MI0028481 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CACUGGACUUGGAGUCAGAAGGCC |
| Species: | Eptesicus fuscus |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000168, mir-378 |
Mature miRNA Sequence: | CACUGGACUUGGAGUCAGAAGGCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Eptesicus fuscus | efu-miR-378 | MIMAT0034813 |