Assay Details
Gene Family ID: | MIPF0000242, mir-425 |
Mature miRNA Sequence: | CCAUCGGGAAUAUCGUGUCCGCCC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Horse | eca-miR-425 | MIMAT0034749 |
Stem-loop Details
Horse
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | eca-miR-425 |
|---|---|
| miRBase Accession Number: | MI0028397 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCAUCGGGAAUAUCGUGUCCGCCC |
| Species: | Horse |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000242, mir-425 |
Mature miRNA Sequence: | CCAUCGGGAAUAUCGUGUCCGCCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Horse | eca-miR-425 | MIMAT0034749 |