Assay Details
Mature miRNA Sequence: | UCGAGGAGCUCACAGUCUAGUA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Dasypus novemcinctus | dno-miR-151-5p | MIMAT0047677 |
| Guinea pig | cpo-miR-151-5p | MIMAT0047030 |
Stem-loop Details
Dasypus novemcinctus
Guinea pig

Genomic Map