Assay Details
Amplicon Length: | 54 |
Chromosome Location: | Chr. 6 - 29022416 - 29022416 [-] on 29022416 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-129-5p | MIMAT0000209 |
Mouse | mmu-miR-129-1-3p | MIMAT0016994 |
Stem-loop Details
Mouse
