Assay Details
| Amplicon Length: | 74 | 
| Chromosome Location: | Chr. 4 - 40723321 - 40723321 [-] on 40723321 Build GRCm38 | 
| Species: | Mouse | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Mouse | mmu-miR-207 | MIMAT0000240 | 
Stem-loop Details
Mouse
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top Ambion® Anti-miR™ miRNA Inhibitor :
						Ambion® Anti-miR™ miRNA Inhibitor :