Assay Details
Amplicon Length: | 95 |
Chromosome Location: | Chr. 16 - 17123864 - 17123864 [+] on 17123864 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-130b-3p | MIMAT0000387 |
| Mouse | mmu-miR-130b-5p | MIMAT0004583 |
Stem-loop Details
Mouse

Genomic Map