Assay Details
Amplicon Length: | 72 |
Chromosome Location: | Chr. 9 - 41531722 - 41531722 [-] on 41531722 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-100-5p | MIMAT0000655 |
| Mouse | mmu-miR-100-3p | MIMAT0017051 |
Stem-loop Details
Mouse

Genomic Map