Assay Details
Amplicon Length: | 84 |
Chromosome Location: | Chr. 11 - 28764029 - 28764029 [+] on 28764029 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-217-3p | MIMAT0017072 |
| Mouse | mmu-miR-217-5p | MIMAT0000679 |
Stem-loop Details
Mouse

Genomic Map