Assay Details
Amplicon Length: | 92 |
Chromosome Location: | Chr. 12 - 109716775 - 109716775 [+] on 109716775 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-666-5p | MIMAT0003737 |
Mouse | mmu-miR-666-3p | MIMAT0004823 |
Stem-loop Details
Mouse
