Assay Details
Amplicon Length: | 64 |
Chromosome Location: | Chr. 10 - 128085084 - 128085084 [+] on 128085084 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-677-5p | MIMAT0003451 |
| Mouse | mmu-miR-677-3p | MIMAT0017246 |
Stem-loop Details
Mouse

Genomic Map