Assay Details
Amplicon Length: | 84 |
Chromosome Location: | Chr. 13 - 75644844 - 75644844 [+] on 75644844 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-682 | MIMAT0003459 |
Stem-loop Details
Mouse
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amplicon Length: | 84 |
Chromosome Location: | Chr. 13 - 75644844 - 75644844 [+] on 75644844 Build GRCm38 |
Species: | Mouse |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-682 | MIMAT0003459 |