Assay Details
Amplicon Length: | 98 |
Chromosome Location: | Chr. X - 100381395 - 100381395 [+] on 100381395 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-676-5p | MIMAT0003781 |
| Mouse | mmu-miR-676-3p | MIMAT0003782 |
Stem-loop Details
Mouse

Genomic Map