Assay Details
Amplicon Length: | 96 |
Chromosome Location: | Chr. X - 66781057 - 66781057 [-] on 66781057 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-883a-3p | MIMAT0004849 |
| Mouse | mmu-miR-883a-5p | MIMAT0004848 |
Stem-loop Details
Mouse

Genomic Map