Assay Details
Amplicon Length: | 107 |
Chromosome Location: | Chr. X - 66801103 - 66801103 [+] on 66801103 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-878-3p | MIMAT0004933 |
Mouse | mmu-miR-878-5p | MIMAT0004932 |
Stem-loop Details
Mouse
