Assay Details
Amplicon Length: | 82 |
Chromosome Location: | Chr. 13 - 109325143 - 109325143 [+] on 109325143 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-582-3p | MIMAT0005292 |
| Mouse | mmu-miR-582-5p | MIMAT0005291 |
Stem-loop Details
Mouse

Genomic Map