Assay Details
Amplicon Length: | 92 |
Chromosome Location: | Chr. 2 - 10517958 - 10517958 [-] on 10517958 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-669h-3p | MIMAT0005842 |
Mouse | mmu-miR-669h-5p | MIMAT0005841 |
Mouse | mmu-miR-669i | MIMAT0005840 |
Stem-loop Details
Mouse
