Assay Details
Amplicon Length: | 60 |
Chromosome Location: | Chr. 12 - 109716072 - 109716072 [-] on 109716072 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-1193-5p | MIMAT0017329 |
| Mouse | mmu-miR-1193-3p | MIMAT0005851 |
Stem-loop Details
Mouse

Genomic Map