Assay Details
Amplicon Length: | 119 |
Chromosome Location: | Chr. 7 - 29083889 - 29083889 [+] on 29083889 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-1963 | MIMAT0009436 |
Stem-loop Details
Mouse
